Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Metal, 100 Mesh, Powder, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
Packaging:
Amber Glass Bottle
Poly Bottle
7440-36-0
7440-36-0
Grade Reagent
CAS Max % 1
Packaging Poly Bottle
Quantity 500 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.