Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony pentafluoride, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
7783-70-2
F5Sb
216.74
MFCD00011219
antimony pentafluoride,antimony v fluoride,pentafluoroantimony,antimony v pentafluoride,antimony fluoride sbf5,hsdb 442,antimonypentafluoride,pentafluorostibine,pentafluorostiborane
7783-70-2
F5Sb
216.74
MFCD00011219
antimony pentafluoride,antimony v fluoride,pentafluoroantimony,antimony v pentafluoride,antimony fluoride sbf5,hsdb 442,antimonypentafluoride,pentafluorostibine,pentafluorostiborane
C-X bond formation reagent
Assay Percent Range 99+%
Linear Formula SbF5
Antimony (Sb) 55.9% min.
Fieser 05,19; 06,23; 09,2
Merck Index 15, 687
Solubility Information Solubility in water: reacts with water. Other solubilities: forming hf and antimonyoxide,soluble in kf and liquid so2
Formula Weight 216.74
Physical Form Shiny Flakes or Powder
Percent Purity 99+%
Density 2.9900g/mL
Fluoride 43.6% min.
Packaging Plastic bottle
Solubility Solubility in water: reacts with water. Other solubilities: forming hf and antimonyoxide, soluble in kf and liquid so2
Specific Gravity 2.99
Storage Information Product may develop some turbidity or precipitate
Boiling Point 149.4°C
Color White
Melting Point 7.0°C
Quantity 25g
Chemical Name or Material Antimony pentafluoride, 99.5+%
Show More Show Less

GHS H Statement
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 232-021-8

RTECSNumber : CC5800000

TSCA : TSCA