Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Potassium Tartrate, Powder, USP, 99-103%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
28300-74-5
C8H10K2O15Sb2
667.87
WBTCZEPSIIFINA-UHFFFAOYNA-J
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
28300-74-5
C8H10K2O15Sb2
667.87
WBTCZEPSIIFINA-UHFFFAOYNA-J
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2

AN140, 28300-74-5, C8H4K2O12Sb2•3H2O

Spectrum™ Antimony Potassium Tartrate, Powder, USP is also referred to as emetic tartar. It has long been known as a powerful emetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
UN Number UN1551
Arsenic (As) 0.00015
Percent Purity 99 to 103%
Lead (Pb) 2e-005
Loss on Drying 0.027
Packaging Poly Bottle
Quantity 125 g