Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Potassium Tartrate, Powder, USP, 99-103%, Spectrum™ Chemical
SDP

Catalog Number 18600734
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
28300-74-5
C8H10K2O15Sb2
667.87
WBTCZEPSIIFINA-UHFFFAOYNA-J
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
28300-74-5
C8H10K2O15Sb2
667.87
WBTCZEPSIIFINA-UHFFFAOYNA-J
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2

AN140, 28300-74-5, C8H4K2O12Sb2•3H2O

Spectrum™ Chemical Antimony Potassium Tartrate, Powder, USP is also referred to as emetic tartar. It has long been known as a powerful emetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Quantity 500 g
UN Number UN1551
Percent Purity 99 to 103%
Grade USP
Loss on Drying 2.7%
Packaging Poly Bottle
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit