Alfa Aesar™ Antimony rod, 12.7mm (0.500 in.) dia., 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4718120

Quantity 80g

 View more versions of this product

Catalog No. AA4718120

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-36-0
    MDL Number MFCD00134030
    Chemical Name or Material Antimony rod, 12.7mm (0.500 in.) dia.

    Specifications

    Quantity 80g
    Odor Odorless
    MDL Number MFCD00134030
    RTECS Number CC4025000
    Weight ≈2.16cm/20g
    TSCA TSCA
    Physical Form Cast Rod
    CAS 7440-36-0
    Chemical Name or Material Antimony rod, 12.7mm (0.500 in.) dia.
    Assay 99.999%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit