Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Shot, 1.5-5mm (0.06-0.2 in.),, Puratronic™, 99.9999% (Metals basis), Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
7440-36-0
Sb
121.76
MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german
5354495
CHEBI:30304
antimony
[Sb]
7440-36-0
Sb
121.76
MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german
5354495
CHEBI:30304
antimony
[Sb]
Assay Percent Range 99.9999% (Metals basis)
Appearance Solid in various forms
Physical Form Shot, 1.5-5mm (0.06-0.2 in.)
Odor Odorless
Color Silver to Gray
Quantity 5g
Chemical Name or Material Antimony

EINECSNumber : 231-146-5

RTECSNumber : CC4025000

TSCA : Yes

Recommended Storage : Ambient temperatures