Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Trichloride, Crystal, 99%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
10025-91-9
10025-91-9
Grade Reagent
CAS Max % 1
Percent Purity 0.99
Packaging Amber Glass Bottle
Quantity 125 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.