Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Trichloride TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass Bottle

A-317, 10025-91-9

Spectrum™ Antimony Trichloride TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS 10025-91-9
CAS Max % 1
Molecular Formula C22H23Cl4N2O8Sb
UN Number UN2922
InChI Key LIARQHHWPMTMFO-CRPMINNHSA-K
SMILES [Cl-].[Cl-].[Cl-].[Sb+3].CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC(Cl)=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(/N)O)C1=O
IUPAC Name antimony(3+) (2E,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-7-chloro-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione trichloride
Molecular Weight (g/mol) 706.99
Packaging Amber Glass Bottle
Quantity 100 mL