Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Trioxide, 99%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
1309-64-4
1309-64-4
Grade Reagent
CAS Max % 1
Arsenic (As) 0.001
Percent Purity 0.99
Chloride 5e-05
Iron (Fe) 2e-05
Packaging Poly Pail
Quantity 12 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.