Antimony(V) chloride, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 4197830

Quantity 250g

 View more versions of this product

Catalog No. AA4197830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-18-9
    Physical Form Liquid
    Assay Percent Range 99%
    Molecular Formula SbCl5
    Formula Weight 299.02
    MDL Number MFCD00011213
    Synonym Antimony pentachloride
    Chemical Name or Material Antimony(V) chloride

    Specifications

    Formula Weight 299.02
    Quantity 250g
    Color Red-yellow
    DOT Information DOT Class: 8, Packaging Group: II
    EINECS Number 231-601-8
    Physical Form Liquid
    Odor Pungent
    Assay Percent Range 99%
    Molecular Formula SbCl5
    Appearance Red-yellow liquid
    MDL Number MFCD00011213
    CAS 7647-18-9
    Synonym Antimony pentachloride
    Health Hazard 1 H314-H411
    Chemical Name or Material Antimony(V) chloride

    H314-H411

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit