Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(V)-chloride, 99%, anhydrous, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
7647-18-9
Cl5Sb
299.01
MFCD00011213
VMPVEPPRYRXYNP-UHFFFAOYSA-I
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
24294
pentachloro-$l^{5}-stibane
Cl[Sb](Cl)(Cl)(Cl)Cl
7647-18-9
Cl5Sb
299.01
MFCD00011213
VMPVEPPRYRXYNP-UHFFFAOYSA-I
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
24294
pentachloro-$l^{5}-stibane
Cl[Sb](Cl)(Cl)(Cl)Cl
C-X bond formation reagent
Assay Percent Range 98.5% min
Linear Formula SbCl5
Fieser 01,42; 04,21; 05,18; 06,22; 07,12; 15,14; 17,17
Merck Index 15, 686
Solubility Information Solubility in water: reacts with water forming hcl and antimonyoxide. Other solubilities: soluble in hcl,carbon disulfide,dichloromethane,soluble in an aqueous solution of tartaric acid,,ccl4 and chloroform
Formula Weight 299.01
Physical Form Liquid
Percent Purity 99%
Density 2.3600g/mL
Packaging Glass bottle
Solubility Solubility in water: reacts with water forming hcl and antimonyoxide. Other solubilities: soluble in hcl, carbon disulfide, dichloromethane, soluble in an aqueous solution of tartaric acid,, ccl4 and chloroform
Specific Gravity 2.36
Storage Information Product may develop some turbidity or precipitate
Boiling Point 92.0°C (30.0 mmHg)
Color Colorless to Yellow
Melting Point 2.8°C
Quantity 1kg
Chemical Name or Material Antimony(V)-chloride, 99%
Show More Show Less

GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 231-601-8

RTECSNumber : CC5075000

TSCA : TSCA