Learn More
Antimycin A, Thermo Scientific Chemicals
Description
Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.
Solubility
Soluble in 100% ethanol. Insoluble in water.
Notes
Isolated from Streptomyces sp.
Specifications
Type | Antimycin A |
Physical Form | Powder |
Melting Point | 149°C to 150°C |
UN Number | UN3462 |
Merck Index | 14,714 |
Solubility Information | Soluble in 100% ethanol. Insoluble in water. |
Formula Weight | 548.63 |
Color | White to Yellow |
Quantity | 5 mg |
Sensitivity | Light sensitive |
Show More |
Safety and Handling
Hazard Category | H300+H310 |
Hazard Statement | GHS H Statement H300 Fatal if swallowed. |
Precautionary Statement | P262-P264b-P270-P280-P301+P310-P302+P350-P310-P330-P361-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S. |
RTECSNumber | CD0350000 |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.