Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antipyrine, MilliporeSigma™ Supelco™

$12.73 - $114.40

Specifications

Grade Analytical Standard
Linear Formula C11H12N2 O
Physical Form Neat
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-4868
View Documents
MilliporeSigma Supelco
A588225G
25 g
Each for $12.73
Add to cart
 
11-101-4869
View Documents
MilliporeSigma Supelco
A5882100G
100 g
Each for $38.05
Add to cart
 
11-101-4867
View Documents
MilliporeSigma Supelco
A5882500G
500 g
Each for $114.40
Add to cart
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

Analytical Standard
C11H12N2 O
Neat
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : CD2450000

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit