Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antipyrine, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
1 kg
10 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
60-80-0
C11H12N2O
188.23
VEQOALNAAJBPNY-UHFFFAOYSA-N
1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
CN1N(C(=O)C=C1C)C1=CC=CC=C1
60-80-0
C11H12N2O
188.23
VEQOALNAAJBPNY-UHFFFAOYSA-N
1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
CN1N(C(=O)C=C1C)C1=CC=CC=C1

A1997, 60-80-0, C11H12N2O

Spectrum™ Antipyrine, USP is a nonsteroidal anti-inflammatory analgesic and antipyretic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Ignition Residue 0.0015
Loss on Drying 0.01
Quantity 10 kg
CAS Max % 1
Percent Purity 98 to 102%