Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antipyrine, USP, 98-102%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
100 g
500 g
1 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
60-80-0
C11H12N2O
188.23
VEQOALNAAJBPNY-UHFFFAOYSA-N
1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
CN1N(C(=O)C=C1C)C1=CC=CC=C1
60-80-0
C11H12N2O
188.23
VEQOALNAAJBPNY-UHFFFAOYSA-N
1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
CN1N(C(=O)C=C1C)C1=CC=CC=C1

A1997, 60-80-0, C11H12N2O

Spectrum™ Chemical Antipyrine, USP is a nonsteroidal anti-inflammatory analgesic and antipyretic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.15%
Loss on Drying 1%
Packaging Poly Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade USP