Learn More
Abnova™ ANTXR2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00118429T01
Catalog No. 89-935-137
Add to cart
Description
Description
Specifications
Specifications
293T
NM_058172.3
pCMV-ANTXR2 full-length
100 μL
Human
ANTXR2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 53.5kDa
Transient overexpression cell lysate was tested with anti-ANTXR2 antibody (Mfr. No. H00118429-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot