Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANXA1 Mouse anti-Human, Unconjugated, Clone: 6E4/3, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB21050
Description
ANXA1 Monoclonal antibody specifically detects ANXA1 in Human samples. It is validated for IHC-P.Specifications
ANXA1 | |
6E4/3 | |
annexin A1 | |
Liquid | |
Mouse | |
IgG1 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
ANXA1 | |
ANX1, LPC1 | |
Mixture of native proteins from human bone tissues | |
100 μg | |
Primary | |
301 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title