Learn More
Abnova™ ANXA4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000307T02
Catalog No. 89-935-159
Add to cart
Description
Description
Specifications
Specifications
293T
NM_001153.2
pCMV-ANXA4 full-length
100 μL
Human
ANXA4 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 36.1kDa
Transient overexpression cell lysate was tested with anti-ANXA4 antibody (Mfr. No. H00000307-D01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot