Learn More
ANXA9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154161AP
Catalog No. 50-173-0211
Add to cart
Description
Description
The annexins include ANXA9 coded protein, which are a family of calcium-dependent phospholipid-binding proteins. Members of the annexin family contain 4 internal repeat domains, each of which includes a type II calcium-binding site. The calcium-binding sites are required for annexins to aggregate and cooperatively bind anionic phospholipids and extracellular matrix proteins. Annexin A9 is a divergent member of the annexin protein family in which all four homologous type II calcium-binding sites in the conserved tetrad core contain amino acid substitutions that ablate their function. However, structural analysis suggests that the conserved putative ion channel formed by the tetrad core is intact.
Specifications
Specifications
ANXA9
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O76027, Q9JHQ0
Annexin 31, Annexin 9, annexin A9, Annexin XXXI, ANX31, ANXA9, Pemphaxin
Rabbit
IgG
150 μL
-20°C
Primary
71790, 8416
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ANXA9
Liquid
Anxa9
ANXA9 Fusion Protein Ag7702
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse