Learn More
Abnova™ AOAH 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000313T01
Catalog No. 89-935-178
Add to cart
Description
Description
Specifications
Specifications
293T
NM_001637.1
pCMV-AOAH full-length
100 μL
Human
AOAH 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 63.36kDa
Transient overexpression cell lysate was tested with anti-AOAH antibody (Mfr. No. H00000313-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot