Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AOAH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA555390
Description
Immunogen sequence: VKKSPILKYS RSGSDICSLP VLAKICQKIK LAMEQSVPFK DVDSDKYSVF PTLRGYHWRG RDCNDSDESV YPGRRPNNWD VHQDSNCNGI WGVDPKDGVP YEKKFCEGSQ PRGIILLGDS AGAH Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
AOAH Polyclonal antibody specifically detects AOAH in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
AOAH | |
Polyclonal | |
Unconjugated | |
AOAH | |
4930433E13Rik; Acyloxyacyl hydrolase; acyloxyacyl hydrolase (neutrophil); Acyloxyacyl hydrolase large subunit; Acyloxyacyl hydrolase small subunit; AOAH; RGD1560514 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
313 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P28039 | |
AOAH | |
Recombinant protein corresponding to Human AOAH | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?
Provide Content Correction