Learn More
Azzota Corp AP-26113 DFS Item
Manufacturer:  Azzota Corp C1031
AP-26113 (Brigatinib) is an orally active, potent and selective dual ALK/EGFR inhibitor. AP-26113 binds to and inhibits ALK kinase and ALK fusion proteins as well as EGFR and mutant forms. Please request if need more quantities
Catalog No. 50-161-3050