Learn More
Abnova™ AP1M2 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00010053T01
Catalog No. 89-935-200
Add to cart
Description
Description
Specifications
Specifications
293T
NM_005498
pCMV-AP1M2 full-length
100 μL
Human
AP1M2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 46.64kDa
Transient overexpression cell lysate was tested with anti-AP1M2 antibody (Mfr. No. H00010053-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot