Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2A2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen AP2A2
Immunogen Synthetic peptides corresponding to AP2A2 (adaptor-related protein complex 2, alpha 2 subunit) The peptide sequence was selected from the C terminal of AP2A2 (NP_036437). Peptide sequence LAKLKKKKGPSTVTDLEDTKRDRSVDVNGGPEPAPASTSAVSTPSPSADL.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16894820 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16894820UL
20ul Each for $149.00
Add to cart
 
NBP168948 View Documents Novus BiologicalsSupplier Diversity Partner
NBP168948
100 ul Each for $329.00
Add to cart
 
Description

Description

AP2A2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

AP2A2
Rabbit
Polyclonal
Unconjugated
161
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to AP2A2 (adaptor-related protein complex 2, alpha 2 subunit) The peptide sequence was selected from the C terminal of AP2A2 (NP_036437). Peptide sequence LAKLKKKKGPSTVTDLEDTKRDRSVDVNGGPEPAPASTSAVSTPSPSADL.
Primary
IgG
100 kDa coated vesicle protein C, Adapter-related protein complex 2 alpha-2 subunit, adaptin, alpha B, Adaptor protein complex AP-2 subunit alpha-2, adaptor-related protein complex 2, alpha 2 subunit, alpha2-adaptin, Alpha-adaptin C, alpha-adaptin C; Huntingtin interacting protein J, AP-2 complex subunit alpha-2, Clathrin assembly protein complex 2 alpha-C large chain, clathrin-associated/assembly/adaptor protein, large, alpha 2, DKFZP564D1864, HIP9ADTAB, Huntingtin yeast partner J, Huntingtin-interacting protein 9, Huntingtin-interacting protein J, HYPJCLAPA2, KIAA0899HIP-9, Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit