Learn More
Aviva Systems Biology Corp AP2B1 Antibody - middle region (ARP53578_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AP2B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AP2B1. Alternate names: ADTB2, AP105B, AP2-BETA, CLAPB1, DKFZp781K0743ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61056