Learn More
Abnova™ AP3S1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00001176T03
Catalog No. 89-935-222
Add to cart
Description
Description
Specifications
Specifications
293T
NM_001284
pCMV-AP3S1 full-length
100 μL
Human
AP3S1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 21.7kDa
Transient overexpression cell lysate was tested with anti-AP3S1 antibody (Mfr. No. H00001176-B02P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot