Learn More
Aviva Systems Biology Corp AP3S1 Antibody - N-terminal region (ARP51935_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51935_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AP3S1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AP3S1. Alternate names: CLAPS3, Sigma3AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60649