Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP3S1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
$152.22 - $436.00
Specifications
Antigen | AP3S1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15544920
|
Novus Biologicals
NBP15544920UL |
20 μL |
Each for $152.22
|
|
NBP155449
|
Novus Biologicals
NBP155449 |
100 μL |
Each for $436.00
|
|
Description
AP3S1 Polyclonal specifically detects AP3S1 in Human samples. It is validated for Western Blot.Specifications
AP3S1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Adapter-related protein complex 3 sigma-1 subunit, adaptor-related protein complex 3, sigma 1 subunit, AP-3 complex sigma-3A subunit, AP-3 complex subunit sigma-3A, CLAPS3AP-3 complex subunit sigma-1, Clathrin-associated/assembly/adapter protein, small 3, clathrin-associated/assembly/adaptor protein, small 3 (22kD), Sigma3A, sigma-3A-adaptin, Sigma3A-adaptin, Sigma-adaptin 3a | |
AP3S1 | |
IgG | |
Affinity Purified | |
22 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q92572 | |
1176 | |
Synthetic peptides corresponding to AP3S1(adaptor-related protein complex 3, sigma 1 subunit) The peptide sequence was selected from the N terminal of AP3S1 (NP_001275). Peptide sequence IKAILIFNNHGKPRLSKFYQPYSEDTQQQIIRETFHLVSKRDENVCNFLE. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title