Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302760A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Transcription factors of the basic helix-loop-helix-zipper(bHLH-ZIP) family contain a basic domain, which is used for DNAbinding, and HLH and ZIP domains, which are used foroligomerization. Transcription factor AP4 activates both viral andcellular genes by binding to the symmetrical DNA sequence CAGCTG.Specifications
AP4 | |
Polyclonal | |
Unconjugated | |
TFAP4 | |
AP-4, bHLHc41, activating enhancer-binding protein 4, class C basic helix-loop-helix protein 41, transcription factor AP-4, transcription factor AP-4 (activating enhancer-binding protein 4) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7023 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q01664 | |
TFAP4 | |
Between 50 and 100 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title