Learn More
APAF1 Rabbit anti-Human, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 217101AP
Catalog No. 50-173-3558
Add to cart
Description
Description
This gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.
Specifications
Specifications
APAF1
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14727, Q9EPV5
APAF 1, APAF1, CED4, KIAA0413
Rabbit
IgG
150 μL
-20°C
Primary
317, 78963
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
APAF1
Liquid
APAF1
APAF1 Fusion Protein Ag16336
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat