Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBB3 Mouse anti-Human, Clone: OTI5G3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503752

Catalog No. 50-167-4569


Add to cart

Description

Description

The protein encoded by this gene is a member of the APBB protein family. It is found in the cytoplasm and binds to the intracellular domain of the Alzheimer ft.s disease beta-amyloid precursor protein (APP) as well as to other APP-like proteins. It is thought that the protein encoded by this gene may modulate the internalization of APP. Multiple transcript variants encoding several different isoforms have been found for this gene.
Specifications

Specifications

APBB3
Monoclonal
0.39 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O95704
FE65L2, SRA
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10307
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI5G3
Unconjugated
APBB3
Liquid
APBB3
Full length human recombit protein of human APBB3 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit