Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBB3 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI5G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271585JF549

Catalog No. N271585J549


Add to cart

Description

Description

APBB3 Monoclonal antibody specifically detects APBB3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

APBB3
OTI5G3
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
APBB3
Full length recombinant protein of human APBB3 (NP_573419) produced in HEK293T cells.
Immunogen affinity purified
Signal Transduction
Primary
10307.0
Flow Cytometry, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
amyloid beta (A4) precursor protein-binding, family B, member 3, amyloid precursor interacting protein, Fe65L2, FE65L2amyloid beta A4 precursor protein-binding family B member 3, FE65-like protein 2, MGC150555, MGC87674, Protein Fe65-like 2, SRA
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit