Learn More
Abnova™ APC 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000324T02
Catalog No. 89-935-254
Add to cart
Description
Description
Specifications
Specifications
293T
BC111591
pCMV-APC full-length
100 μL
Human
APC 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 312.73kDa
Transient overexpression cell lysate was tested with anti-APC antibody (Mfr. No. H00000324-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot