Learn More
BiolegendSupplier Diversity Partner APC anti-human CD158e1 (KIR3DL1, NKB1)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 312716
APC anti-human CD158e1 (KIR3DL1, NKB1) [DX9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-012