Learn More
Biolegend APC anti-human IgE
Supplier:  Biolegend 325507
Encompass_Preferred
APC anti-human IgE [MHE-18]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-285