Learn More
BiolegendSupplier Diversity Partner APC/Cyanine7 anti-human CD11b  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 301342
APC/Cyanine7 anti-human CD11b [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 tests
Catalog No. 50-711-970