Learn More
Biolegend APC/Cyanine7 anti-human CD279 (PD-1)
Supplier:  Biolegend 329922
Encompass_Preferred
APC/Cyanine7 anti-human CD279 (PD-1) [EH12.2H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. 50-167-532