Learn More
Biolegend APC/Cyanine7 Mouse IgG2a, κ isotype Ctrl
Supplier:  Biolegend 400230
Encompass_Preferred
APC/Cyanine7 Mouse IgG2a, κ isotype Ctrl [MOPC-173]; Isotype: Mouse IgG2a, κ; Apps: FC, ICFC; Size: 100 tests
Catalog No. 50-168-873