Learn More
Aat BioquestSupplier Diversity Partner ReadiUse™ Preactivated APC-iFluor™ 700 Tandem  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2570
Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga. Like other phycobiliproteins, APC is fluorescent, with an extremely high absorptivity and a high quantum efficiency.
Catalog No. 50-195-3190