Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC Mouse anti-Human, Clone: ALi 12-28, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30717910UG

 View more versions of this product

Catalog No. NB121831


Add to cart

Description

Description

APC Monoclonal antibody specifically detects APC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications

Specifications

APC
ALi 12-28
Unconjugated
10 mM PBS with 0.05% BSA
adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC
Recombinant fragment of human APC protein (around aa 1-433)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
0.2 mg/ml
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway
Primary
324
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit