Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APC-PCI Complex Mouse anti-Human, Alexa Fluor™ 350, Clone: M36, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241380AF350
Description
APC-PCI Complex Monoclonal specifically detects APC-PCI Complex in Human samples. It is validated for ELISA.Specifications
APC-PCI Complex | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Mixture of PCI in complex with PSA (approx. 80%) and PCI cleaved from such a complex (approx. 20%). | |
100 μL | |
Antibody is specific for APC:PCI complex and cleaved PCI but has little reactivity towards native PCI. The dissociation constants (as determined by standard surface plasmon resonance technique) are: Kd= 4x10e-10 M for PCI in complex with APC and 2x10e-10 M for cleaved PCI. The value of Kd for native PCI is too low to measure, i.e. > 10e-5 M. | |
Store at 4°C in the dark. |
ELISA | |
M36 | |
ELISA | |
Mouse | |
Protein A or G purified | |
Primary | |
Human | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title