Learn More
APC11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150651AP
Catalog No. 50-172-9946
Add to cart
Description
Description
Component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. The APC/C complex acts by mediating ubiquitination and subsequent degradation of target proteins: it mainly mediates the formation of  ft.Lys-11 ft.-linked polyubiquitin chains and, to a lower extent, the formation of  ft.Lys-48 ft.- and  ft.Lys-63 ft.-linked polyubiquitin chains. May recruit the E2 ubiquitin-conjugating enzymes to the complex.
Specifications
Specifications
APC11
0.47 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9CPX9, Q9NYG5
ANAPC11, APC11, Apc11p, Cyclosome subunit 11, HSPC214
Rabbit
IgG
150 μL
-20°C
Primary
51529, 66156
Western Blot
Unconjugated
ANAPC11
Liquid
ANAPC11
APC11 Fusion Protein Ag7069
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse