Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apc5 Antibody (APC5#4), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250106UV
Description
Apc5 Monoclonal specifically detects Apc5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Apc5 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ANAPC5 | |
GST-human APC5 | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair | |
51433 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
APC5#4 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
anaphase promoting complex subunit 5, anaphase-promoting complex subunit 5, APC5Cyclosome subunit 5 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction