Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APCDD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548559

Catalog No. PA548559

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with xenopus based on sequence homology.

APCDD1 (Drapc1) is a novel protein that has been shown to be a target of Wnt/beta-catenin signaling pathway in cancer cell lines. APCDD1 is overexpressed in colorectal carcinogenesis and is deregulated in CTNNB1-mutated Wilms tumors.
Specifications

Specifications

APCDD1
Polyclonal
Unconjugated
APCDD1
Liquid
APCDD1
KLH conjugated synthetic peptide between 69-98 amino acids from the N-terminal region of human APCDD1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 0.09% sodium azide
Q8J025
Protein APCDD1, Adenomatosis polyposis coli down-regulated 1 protein, APCDD1 (HGNC:15718)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
147495
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit