Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Apcin
Cdc20 inhibitor; inhibits Cdc20-substrate interaction
$247.08 - $1008.04
Specifications
Chemical Name or Material | 3-(2-Methyl-5-nitroimidazol-1-yl)-N-(2,2,2-trichloro-1-phenylaminoethyl)propionamide |
---|---|
CAS | 300815-04-7 |
Target | Ubiquitin Ligase (E3) Inhibitors |
Molecular Formula | C13H14Cl3N7O4 |
Formula Weight | 438.65 |
Description
Cdc20 inhibitor; inhibits Cdc20-substrate interaction. Blocks substrate-mediated Cdc20 loading onto the anaphase-promoting complex (APC). Inhibits APC-dependent ubiquitylation. Prolongs mitotic duration in RPE1 cells in combination with proTAME (prodrug of TAME (Cat.No. 4506)) in vitro.Specifications
3-(2-Methyl-5-nitroimidazol-1-yl)-N-(2,2,2-trichloro-1-phenylaminoethyl)propionamide | |
Ubiquitin Ligase (E3) Inhibitors | |
438.65 |
300815-04-7 | |
C13H14Cl3N7O4 | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title