Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Apelin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551077
Description
Apelin Polyclonal Antibody detects endogenous levels of Apelin protein.
This gene encodes a peptide that functions as an endogenous ligand for the G protein coupled receptor APJ. The encoded protein is synthesized as a prepropeptide that is processed into biologically active C-terminal fragments. The peptide fragments activate different tissue specific signaling pathways that regulate diverse biological functions including fluid homeostasis, cardiovascular function and insulin secretion. This protein also functions as a coreceptor for the human immunodeficiency virus 1.
Specifications
Apelin | |
Polyclonal | |
Unconjugated | |
Apln | |
6030430G11Rik; AGTRL1 ligand; APEL; Apelin; apelin, AGTRL1 ligand; Apelin-13; Apelin-28; Apelin-31; Apelin-36; APJ endogenous ligand; APLN; preproapelin; XNPEP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30878, 58812, 8862 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9R0R3, Q9R0R4, Q9ULZ1 | |
Apln | |
Synthesized peptide derived from the C-terminal region of human Apelin. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction