Learn More
Peptides InternationalSupplier Diversity Partner APETx2 (0.1 mg vial), CAS# 713544-47-9, MF= C196H280N54O61S6 DFS Item
Manufacturer:  Peptides InternationalSupplier Diversity Partner APE4472S1EA
Gly-Thr-Ala-Cys-Ser-Cys-Gly-Asn-Ser-Lys-Gly-Ile-Tyr-Trp-Phe-Tyr-Arg-Pro-Ser-Cys-Pro-Thr-Asp-Arg-Gly-Tyr-Thr-Gly-Ser-Cys-Arg-Tyr-Phe-Leu-Gly-Thr-Cys-Cys-Thr-Pro-Ala-Asp, Application: Selective Blocker of Acid-Sensing Ion Channel, ASIC3, origin/species: Sea Anemone, Anthopleura elegantissima (Synthetic Product), one letter code: GTACSCGNSKGIYWFYRPSCPTDRGYTGSCRYFLGTCCTPAD, MW: 4561, Disulfide bonds: Reported Disulfide Bonds Between Cys4-Cys37, Cys6-Cys30, and Cys20-Cys38
Catalog No. 50-195-2397