Learn More
Molecular Probes™ APEX™ Alexa Fluor™ 488 Antibody Labeling Kit Non-distribution product as customer accommodation.
APEX™ Alexa Fluor™ 488 Antibody Labeling Kit
Manufacturer:  Molecular Probes™ A10468
Catalog No. A10468
Add to cart
Description
Description
The APEX™ Antibody Labeling Kits are our best option for covalently attaching a fluorophore to small amounts of IgG antibody (~10–20 µg). It is ideal for the efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. Labeled antibodies are ready for use in imaging or flow cytometry applications in as little as 2.5 hours with very little hands on time.
Important Features of Alexa Fluor™ APEX™ Antibody Labeling Kits:
  • Labeled antibodies typically ready to use in 2.5 hours (~15 minutes hands on time)
  • Designed to label 10–20 µg of IgG
  • Covalent attachment
  • Compatible with contaminating proteins or stabilizers like BSA
  • No columns needed; everything you need is supplied for 5 separate labelings
  • Choose from Alexa Fluor™ 488, 555, 568, 594, and 647 dyes, Oregon Green™ 488 dye, Pacific Blue™ dye, and Biotin-XX.
    Better Results and Workflows With Primary Labeled Antibodies
    A primary antibody directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Further, multiple primary antibodies of the same isotype or derived from the same species can easily be used in the same experiment if they are directly labeled with compatible fluorophores.
    Contaminating Proteins or Protein Stabilizers Are Not a Problem
    Many IgG antibodies are often available only in small quantities and packaged with stabilizing proteins, such as BSA, or other contaminants which can interfere with the amine-reactive labeling reagents. The APEX™ Antibody Labeling Kits avoids this by utilizing a solid-phase labeling technique that captures the IgG antibody on the resin inside the APEX™ antibody labeling tip. Contaminants are simply eluted through the tip, prior to applying the amine-reactive label.
    Learn More about Protein and Antibody Labeling
    We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.
    We’ll Make a Custom Antibody Conjugate for You
    If you can’t find what you’re looking for in our stocked list, we’ll prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 9001:2000 certified.
    For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
  • Specifications
    Specifications
    APEX™ Alexa Fluor™ 488 Antibody Labeling Kit, Green, Fluorescent Detection Method, Alexa Fluor™ 488 Dye, C27H13Cl2N2Na3O14S3 Molecular Formula, 825.46 Molecular Weight, 495/519nm Excitation/Emission Wavelength, Used for Labeling Chemistr
    Labeling chemistry
    5 Vials reactive fluorescent label (component A), 5 APEX™ antibody labeling tips (component B), 1.8mL wash buffer (Component C), dimethyl sulfoxide (DMSO) (component D), 100μL labeling buffer (component E), 100μL neutralization buffer (compone
    5 rxn(s)
    2 to 6°C
    825.46
    519nm
    Green
    Alexa Fluor™ 488
    5 Vials reactive fluorescent label (component A), 5 APEX™ antibody labeling tips (component B), 1.8mL wash buffer (Component C), dimethyl sulfoxide (DMSO) (component D), 100μL labeling buffer (component E), 100μL neutralization buffer (compone
    APEX™ Alexa Fluor™ 488 Antibody Labeling Kit
    1 Kit(s)
    495nm
    Documents
    Provide Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Cancel Submit
    For Research Use Only. Not for use in diagnostic procedures.