Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aphidicolin, >99%, MP Biomedicals™

Click to view available options
Quantity:
1mg
5mg
10mg
C20H34O4
338.5
MFCD00083214
APC
C20H34O4
338.5
MFCD00083214
APC
  • It is an antibiotic, a potent antiviral and antimitotic agent and DNA polymerase inhibitor.
  • Inhibits the early S-phase of the cell cycle. DNA polymerase α and δ reversible inhibitor.
  • Potentiates apoptosis induced by arabinosyl nucleosides in leukemia cell lines.
  • Also induces apoptosis in HeLa S3 cells, but inhibits vincristine-induced apoptosis in the p53-negative human prostate cancer cell line PC-3.
Quantity 10mg
Color White
Optical Rotation [α]27/D +12°, c = 1 in methanol (Lit.)
Melting Point 227°C to 233°C (literature)
Beilstein 4689958
Solubility Information Soluble in DMSO ethanol (50mg/mL ) and soluble in methanol (10mg/mL - clear, colorless solution).
Physical Form Free Flowing Powder
Grade >98%

RTECSNumber : PB9185000

storageNote1 : Store at -20°C.