Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apicidin, Tocris Bioscience™
Click to view available options
Quantity:
1mg
Description
Specifications
Quantity | 1mg |
Formula Weight | 623.78 |
Percent Purity | >98% |
Chemical Name or Material | Apicidin |
Safety and Handling
Recommended Storage : Store at -20°C